Gene Transcripts

Gene Symbol
Pura
Entrez Gene ID
19290
Gene Name
purine rich element binding protein A
Gene Aliases
CAGER-1, Pur-alpha, ssCRE-BP
Chromosome Location
Chr. 18: 36280551 - 36291721 on Build GRCm38
UniGene ID
Mm.231802
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008989.3 NP_033015.1 2 769
GenBank mRNA AF017631.1 - 1 294
U02098.1 - 2 769

View Full Product Details