Gene Transcripts

Gene Symbol
Swap70
Entrez Gene ID
20947
Gene Name
SWA-70 protein
Gene Aliases
70kDa, AV235546
Chromosome Location
Chr. 7: 110221703 - 110283506 on Build GRCm38
UniGene ID
Mm.334144
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009302.3 NP_033328.3 2 317
GenBank mRNA AF053974.1 - 2 209
AK019882.1 - 2 260
AK041505.1 - 2 265
AK045295.1 - 2 289
AK146773.1 - 2 309
AK172990.1 - 2 254
BC065136.1 - 2 259 30546814
BY118458.1 - 2 318

View Full Product Details