Gene Transcripts

Gene Symbol
Tcf15
Entrez Gene ID
21407
Gene Name
transcription factor 15
Gene Aliases
Meso1, bHLH-EC2
Chromosome Location
Chr. 2: 152143046 - 152149097 on Build GRCm38
UniGene ID
Mm.3881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009328.2 NP_033354.2 2 686
GenBank mRNA BC027533.1 - 2 686
U18658.1 - 2 726

View Full Product Details