Gene Transcripts

Gene Symbol
Ambn
Entrez Gene ID
11698
Gene Name
ameloblastin
Gene Aliases
Chromosome Location
Chr. 5: 88455991 - 88468531 on Build GRCm38
UniGene ID
Mm.8437
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017250.1 - 4 286
BC087927.1 - 4 286 30504437
U65021.1 - 4 266

View Full Product Details