Gene Transcripts

Gene Symbol
C1qb
Entrez Gene ID
12260
Gene Name
complement component 1, q subcomponent, beta polypeptide
Gene Aliases
Chromosome Location
Chr. 4: 136880145 - 136886177 on Build GRCm38
UniGene ID
Mm.2570
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009777.2 NP_033907.1 3 572
GenBank mRNA AK140712.1 - 3 529
AK152764.1 - 3 591
AK153124.1 - 3 464
BC067001.1 - 3 508 5715633
M22531.1 - 3 572
M36293.1 - 3 572
X16874.1 - 2 427

View Full Product Details