Gene Transcripts

Gene Symbol
Padi4
Entrez Gene ID
18602
Gene Name
peptidyl arginine deiminase, type IV
Gene Aliases
Pad4, Pdi4
Chromosome Location
Chr. 4: 140745508 - 140774239 on Build GRCm38
UniGene ID
Mm.250358
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011061.2 NP_035191.2 2 256
GenBank mRNA AB013850.1 - 2 249
AK137625.1 - 2 257

View Full Product Details