Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011171.2 | NP_035301.2 | 3 | 738 | |
GenBank mRNA | AK136541.1 | - | 3 | 599 | |
AK143719.1 | - | 3 | 840 | ||
AK167302.1 | - | 3 | 678 | ||
BC028755.1 | - | 3 | 611 | 1054063 | |
CT010175.1 | - | 3 | 557 | ||
L39017.1 | - | 3 | 612 | ||
X63748.2 | - | 3 | 644 |