Gene Transcripts

Gene Symbol
Tcf12
Entrez Gene ID
21406
Gene Name
transcription factor 12
Gene Aliases
A130037E08Rik, ALF1, HEB, HEBAlt, HTF-4, HTF4, ME1, REB, bHLHb20
Chromosome Location
Chr. 9: 71844252 - 72111819 on Build GRCm38
UniGene ID
Mm.171615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253862.1 NP_001240791.1 6 560
NM_011544.3 NP_035674.2 6 617
GenBank mRNA AK134037.1 - 6 561
AK143246.1 - 6 620
AK169800.1 - 6 646
M97635.1 - 6 588
X64840.1 - 6 417

View Full Product Details