Gene Transcripts
- Gene Symbol
- Tcf12
- Entrez Gene ID
- 21406
- Gene Name
- transcription factor 12
- Gene Aliases
- A130037E08Rik, ALF1, HEB, HEBAlt, HTF-4, HTF4, ME1, REB, bHLHb20
- Chromosome Location
- Chr. 9: 71844252 - 72111819 on Build GRCm38
- UniGene ID
- Mm.171615
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001253862.1 |
NP_001240791.1
|
12, 13 |
1240 |
|
NM_001253863.1 |
NP_001240792.1
|
5, 6 |
619 |
|
NM_001253864.1 |
NP_001240793.1
|
5, 6 |
619 |
|
NM_001253865.1 |
NP_001240794.1
|
7, 8 |
866 |
|
NM_011544.3 |
NP_035674.2
|
12, 13 |
1297 |
|
GenBank mRNA |
AK078415.1 |
- |
5, 6 |
612 |
|
AK132002.1 |
- |
5, 6 |
604 |
|
AK133958.1 |
- |
5, 6 |
607 |
|
AK134037.1 |
- |
12, 13 |
1241 |
|
AK135376.1 |
- |
5, 6 |
587 |
|
BC037097.1 |
- |
7, 8 |
854 |
5345693 |
M97635.1 |
- |
12, 13 |
1268 |
|
X64840.1 |
- |
12, 13 |
1097 |
|
View Full Product Details