Gene Transcripts

Gene Symbol
Zfp54
Entrez Gene ID
22712
Gene Name
zinc finger protein 54
Gene Aliases
2810034D10Rik, KRAB10, Zfp-54, Zfp76, mszf83
Chromosome Location
Chr. 17: 21423227 - 21435640 on Build GRCm38
UniGene ID
Mm.12916
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011760.2 NP_035890.2 4 612
GenBank mRNA AF080070.1 - 4 573
AK148292.1 - 4 605
BC080782.1 - 5 664 30468186
U29509.1 - 2 104

View Full Product Details