Gene Transcripts
- Gene Symbol
- Banf1
- Entrez Gene ID
- 23825
- Gene Name
- barrier to autointegration factor 1
- Gene Aliases
- Baf, Bcrp1, L2bp1/Baf
- Chromosome Location
- Chr. 19: 5364633 - 5366876 on Build GRCm38
- UniGene ID
- Mm.358649
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038231.2 |
NP_001033320.1
|
2, 3 |
248 |
|
NM_001286608.1 |
NP_001273537.1
|
2, 3 |
590 |
|
NM_011793.3 |
NP_035923.1
|
2, 3 |
456 |
|
GenBank mRNA |
AB025349.1 |
- |
2, 3 |
188 |
|
AF037080.1 |
- |
1, 2 |
131 |
|
AK003025.1 |
- |
2, 3 |
232 |
|
AK004212.1 |
- |
2, 3 |
215 |
|
AK010257.1 |
- |
2, 3 |
234 |
|
AK011079.1 |
- |
2, 3 |
237 |
|
AK011100.1 |
- |
2, 3 |
235 |
|
AK012588.1 |
- |
2, 3 |
218 |
|
AK088896.1 |
- |
2, 3 |
212 |
|
BC003709.1 |
- |
2, 3 |
225 |
3599828 |
BY111854.1 |
- |
2, 3 |
321 |
|
CK780850.1 |
- |
2, 3 |
220 |
|
View Full Product Details