Gene Transcripts
- Gene Symbol
- Bpnt1
- Entrez Gene ID
- 23827
- Gene Name
- bisphosphate 3'-nucleotidase 1
- Gene Aliases
- BPntase
- Chromosome Location
- Chr. 1: 185331965 - 185357769 on Build GRCm38
- UniGene ID
- Mm.227549
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011794.3 |
NP_035924.2
|
5, 6 |
502 |
|
GenBank mRNA |
AF125043.1 |
- |
5, 6 |
460 |
|
AK008091.1 |
- |
4, 5 |
355 |
|
AK008714.1 |
- |
5, 6 |
483 |
|
AK093938.1 |
- |
5, 6 |
488 |
|
AK150877.1 |
- |
5, 6 |
482 |
|
AK151931.1 |
- |
5, 6 |
506 |
|
AK152009.1 |
- |
5, 6 |
482 |
|
AK154435.1 |
- |
5, 6 |
482 |
|
BC011036.1 |
- |
5, 6 |
455 |
4160742 |
View Full Product Details