Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013477.3 | NP_038505.2 | 2 | 361 | |
GenBank mRNA | AB088408.1 | - | 2 | 250 | |
AK083372.1 | - | 2 | 362 | ||
AK133921.1 | - | 2 | 369 | ||
AK171515.1 | - | 2 | 331 | ||
BC011075.1 | - | 2 | 252 | 3662404 | |
U13840.1 | - | 2 | 250 | ||
U21549.1 | - | 2 | 233 |