Gene Transcripts

Gene Symbol
Trove2
Entrez Gene ID
20822
Gene Name
TROVE domain family, member 2
Gene Aliases
1810007I17Rik, A530054J02Rik, AI646302, SS-A/Ro, Ssa, Ssa2
Chromosome Location
Chr. 1: 143750790 - 143777051 on Build GRCm38
UniGene ID
Mm.40370
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013835.2 NP_038863.1 2 368
GenBank mRNA AK076167.1 - 2 345
AK167379.1 - 2 309
AK167555.1 - 2 309
AK167645.1 - 2 309
BC040368.1 - 1 239
BC051974.1 - 1 245 5697399
BC052380.1 - 1 245 5697399
L81154.1 - 1 216
U66843.1 - 2 295

View Full Product Details