Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013782.4 | NP_038810.2 | 8, 9 | 913 | |
GenBank mRNA | AF099053.1 | - | 8, 9 | 797 | |
AK036368.1 | - | 8, 9 | 914 | ||
AK048026.1 | - | 8, 9 | 896 | ||
AK081275.1 | - | 8, 9 | 880 | ||
AK137415.1 | - | 874 | |||
BC009013.1 | - | 6, 7 | 580 | ||
BC053517.1 | - | 8, 9 | 850 | 6315175 |