Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013863.5 | NP_038891.4 | 4 | 1334 | |
GenBank mRNA | AB041583.1 | - | 4 | 1301 | |
AF130471.1 | - | 4 | 1067 | ||
AK007414.1 | - | 4 | 1145 | ||
AK016510.1 | - | 4 | 1141 | ||
AK036675.1 | - | 4 | 1143 | ||
AK171734.1 | - | 4 | 1276 | ||
AK191626.1 | - | 1 | 60 | ||
BC145765.1 | - | 4 | 1247 | 40131068 |