Gene Transcripts

Gene Symbol
Grk4
Entrez Gene ID
14772
Gene Name
G protein-coupled receptor kinase 4
Gene Aliases
A830025H08Rik, GRK, Gprk2l, Gprk4
Chromosome Location
Chr. 5: 34660287 - 34755305 on Build GRCm38
UniGene ID
Mm.117076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080743.1 NP_001074212.1 3 815
NM_019497.2 NP_062370.2 3 815
GenBank mRNA AF040745.1 - 3 241
AK020792.1 - 3 815
AK078475.1 - 3 687
AK132685.1 - 3 816
BC150691.1 - 3 339 9087602

View Full Product Details