Gene Transcripts

Gene Symbol
KCNMB1
Entrez Gene ID
3779
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 1
Gene Aliases
BKbeta1, K(VCA)beta, SLO-BETA, hbeta1, hslo-beta, k(VCA)beta-1, slo-beta-1
Chromosome Location
Chr. 5: 170378161 - 170389677 on Build GRCh38
UniGene ID
Hs.484099
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004137.3 NP_004128.1 4 1168
GenBank mRNA U25138.1 - 4 884
U42600.1 - 4 778
U61536.1 - 4 830

View Full Product Details