Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019673.2 | NP_062647.2 | 4 | 618 | |
GenBank mRNA | AF041476.1 | - | 4 | 438 | |
AK002724.1 | - | 4 | 414 | ||
AK090154.1 | - | 4 | 437 | ||
AK167905.1 | - | 4 | 432 | ||
AK169278.1 | - | 4 | 437 | ||
BC001994.1 | - | 4 | 406 | 3491205 | |
BC094498.1 | - | 4 | 434 | 4209491 | |
CF725603.1 | - | 4 | 618 |