Gene Transcripts
- Gene Symbol
- Adrm1
- Entrez Gene ID
- 56436
- Gene Name
- adhesion regulating molecule 1
- Gene Aliases
- 1110063P18Rik, 2510006J17Rik, AA408205, ARM-1, AU043535, Arm1, Gp110, Rpn13
- Chromosome Location
- Chr. 2: 180171490 - 180176286 on Build GRCm38
- UniGene ID
- Mm.316654
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019822.3 |
NP_062796.2
|
2, 3 |
320 |
|
GenBank mRNA |
AF225959.1 |
- |
2, 3 |
236 |
|
AK010919.1 |
- |
2, 3 |
297 |
|
AK082084.1 |
- |
2, 3 |
303 |
|
AK143843.1 |
- |
2, 3 |
295 |
|
AK145474.1 |
- |
2, 3 |
282 |
|
AK145792.1 |
- |
2, 3 |
311 |
|
AK150091.1 |
- |
2, 3 |
273 |
|
AK166878.1 |
- |
2, 3 |
309 |
|
AK207110.1 |
- |
1, 2 |
140 |
|
BC008974.1 |
- |
2, 3 |
320 |
3586698 |
BC019746.1 |
- |
2, 3 |
249 |
4188750 |
BC031517.1 |
- |
2, 3 |
263 |
4008987 |
View Full Product Details