Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080965.1 | NP_001074434.1 | 3 | 318 | |
NM_001080966.1 | NP_001074435.1 | 3 | 604 | ||
NM_020572.2 | NP_065597.2 | 3 | 482 | ||
GenBank mRNA | AF054620.1 | - | 3 | 569 | |
BC064780.1 | - | 3 | 486 | 30476803 | |
BC100337.1 | - | 3 | 239 | ||
BG080448.2 | - | 3 | 295 | ||
DV648982.1 | - | 3 | 346 |