Gene Transcripts

Gene Symbol
Anapc5
Entrez Gene ID
59008
Gene Name
anaphase-promoting complex subunit 5
Gene Aliases
2510006G12Rik, AA408751, AA536819, AA986414, APC5, Anpc5
Chromosome Location
Chr. 5: 122787459 - 122821342 on Build GRCm38
UniGene ID
Mm.332667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042491.2 NP_001035956.1 1, 2 371
NM_001289517.1 NP_001276446.1 1, 2 371
NM_001289518.1 NP_001276447.1 1, 2 371
NM_001289519.1 NP_001276448.1 1, 2 371
NM_001289520.1 NP_001276449.1 1, 2 371
NM_021505.3 NP_067480.2 1, 2 371
GenBank mRNA AB041593.1 - 1, 2 314
AK013978.1 - 1, 2 295
AK031432.1 - 1, 2 296
AK049719.1 - 1, 2 325
AK075969.1 - 1, 2 310
AK088327.1 - 1, 2 334
AK147951.1 - 1, 2 310
AK148679.1 - 1, 2 326
AK151314.1 - 1, 2 317
AK151905.1 - 1, 2 317
AK152528.1 - 1, 2 287
AK152981.1 - 1, 2 317
AK153373.1 - 1, 2 317
AK153422.1 - 1, 2 317
AK159204.1 - 1, 2 317
AK159713.1 - 1, 2 310
AK159916.1 - 1, 2 310
AK167561.1 - 1, 2 310
AK168135.1 - 1, 2 295
AK168737.1 - 1, 2 311
AK168814.1 - 1, 2 310
AK169196.1 - 1, 2 334
AK180837.1 - 1, 2 44
AK182300.1 - 1, 2 44
AK186196.1 - 1, 2 49
AK192754.1 - 1, 2 67
AK194108.1 - 1, 2 36
AK203216.1 - 20
AK215079.1 - 1, 2 44
BC010339.1 - 1, 2 287 2648173
BC046566.1 - 1, 2 350 2650790
BC046804.1 - 1, 2 308 6404923
BQ950394.1 - 1, 2 371

View Full Product Details