Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021509.5 | NP_067484.2 | 1, 2 | 359 | |
GenBank mRNA | AB041606.1 | - | 1, 2 | 294 | |
AK014340.1 | - | 1, 2 | 360 | ||
AK030274.1 | - | 1, 2 | 300 | ||
AK081586.1 | - | 1, 2 | 325 | ||
AK162030.1 | - | 1, 2 | 301 | ||
BC025892.1 | - | 1, 2 | 57 | ||
BC037684.1 | - | 1, 2 | 297 | ||
BC057652.1 | - | 1, 2 | 273 | 4017199 |