Gene Transcripts
- Gene Symbol
- Akr1a1
- Entrez Gene ID
- 58810
- Gene Name
- aldo-keto reductase family 1, member A1 (aldehyde reductase)
- Gene Aliases
- 2610201A18Rik, Akr1a4
- Chromosome Location
- Chr. 4: 116636510 - 116651674 on Build GRCm38
- UniGene ID
- Mm.30085
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021473.3 |
NP_067448.1
|
7, 8 |
1175 |
|
GenBank mRNA |
AF225564.1 |
- |
7, 8 |
1080 |
|
AK005162.1 |
- |
4, 5 |
616 |
|
AK005376.1 |
- |
7, 8 |
1110 |
|
AK009462.1 |
- |
7, 8 |
952 |
|
AK011157.1 |
- |
4, 5 |
616 |
|
AK011209.1 |
- |
4, 5 |
616 |
|
AK011221.1 |
- |
4, 5 |
615 |
|
AK011321.1 |
- |
7, 8 |
1104 |
|
AK011388.1 |
- |
7, 8 |
1130 |
|
AK011667.1 |
- |
7, 8 |
1131 |
|
AK011794.1 |
- |
7, 8 |
1104 |
|
AK011856.1 |
- |
7, 8 |
1131 |
|
AK011906.1 |
- |
7, 8 |
1109 |
|
AK011908.1 |
- |
7, 8 |
1144 |
|
AK011918.1 |
- |
7, 8 |
1130 |
|
AK128906.1 |
- |
7, 8 |
897 |
|
AK144198.1 |
- |
7, 8 |
1131 |
|
AK146081.1 |
- |
7, 8 |
1129 |
|
AK146278.1 |
- |
7, 8 |
1131 |
|
AK152086.1 |
- |
7, 8 |
1110 |
|
AK159573.1 |
- |
7, 8 |
1127 |
|
AK164461.1 |
- |
7, 8 |
1130 |
|
AK168950.1 |
- |
7, 8 |
1126 |
|
AK181349.1 |
- |
3, 4 |
350 |
|
AY207463.1 |
- |
7, 8 |
1152 |
|
BC003850.1 |
- |
7, 8 |
958 |
|
BC039926.1 |
- |
7, 8 |
1124 |
3156658 |
BC046762.1 |
- |
7, 8 |
1081 |
5693736 |
View Full Product Details