Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022988.2 | NP_075364.2 | 1 | 128 | |
GenBank mRNA | AF284439.1 | - | 2 | 605 | |
AK003978.1 | - | 2 | 235 | ||
AK011670.1 | - | 2 | 145 | ||
AK076788.1 | - | 241 | |||
AK131631.1 | - | 2 | 491 | ||
AK154815.1 | - | 2 | 220 | ||
AK158616.1 | - | 2 | 511 | ||
BC065163.1 | - | 1 | 128 | 6404016 | |
BC089475.1 | - | 2 | 148 | 30504614 |