Gene Transcripts

Gene Symbol
Nkg7
Entrez Gene ID
72310
Gene Name
natural killer cell group 7 sequence
Gene Aliases
2500004F03Rik
Chromosome Location
Chr. 7: 43437138 - 43438246 on Build GRCm38
UniGene ID
Mm.34613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024253.4 NP_077215.2 1, 2 331
GenBank mRNA AK010872.1 - 1, 2 331
AK088383.1 - 1, 2 340
BC055882.1 - 1, 2 396 3497559

View Full Product Details