Gene Transcripts

Gene Symbol
Aggf1
Entrez Gene ID
66549
Gene Name
angiogenic factor with G patch and FHA domains 1
Gene Aliases
2010009L17Rik, 2310029P06Rik, AW112072, Peg3, VG5Q
Chromosome Location
Chr. 13: 95350683 - 95375357 on Build GRCm38
UniGene ID
Mm.36785
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025630.3 NP_079906.2 14 3173
GenBank mRNA AK009533.1 - 10 2136
AY500995.1 - 14 3146
BC027286.1 - 9 1848
BC028451.1 - 1 1308
BC052410.1 - 14 3150 5716440

Gene Symbol
Gm12503
Entrez Gene ID
100040313
Gene Name
predicted gene 12503
Gene Aliases
OTTMUSG00000007207
Chromosome Location
Chr. 4: 44092636 - 44118080 on Build GRCm38
UniGene ID
Mm.301781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK035708.1 - 2 3080

View Full Product Details