Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025988.2 | NP_080264.1 | 6 | 885 | |
GenBank mRNA | AK008170.1 | - | 5 | 418 | |
AK008243.1 | - | 4 | 434 | ||
AK079604.1 | - | 5 | 754 | ||
AK148438.1 | - | 380 | |||
BC048371.1 | - | 5 | 799 | 6396076 | |
BI734653.1 | - | 5 | 419 |