Gene Transcripts

Gene Symbol
Ppp2r2d
Entrez Gene ID
52432
Gene Name
protein phosphatase 2, regulatory subunit B, delta
Gene Aliases
1300017E19Rik, D7Ertd753e, MDS026
Chromosome Location
Chr. 7: 138846386 - 138883057 on Build GRCm38
UniGene ID
Mm.258739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026391.2 NP_080667.1 7 818
GenBank mRNA AF366393.1 - 7 726
AK005033.1 - 6 707
AK129386.2 - 7 845
BC066022.1 - 7 818 6828707

View Full Product Details