Gene Transcripts
- Gene Symbol
- Wdr77
- Entrez Gene ID
- 70465
- Gene Name
- WD repeat domain 77
- Gene Aliases
- 2610003I18Rik, 2610312E17Rik, C79984, p44/MEP50
- Chromosome Location
- Chr. 3: 105959498 - 105969760 on Build GRCm38
- UniGene ID
- Mm.5110
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_027432.3 |
NP_081708.1
|
2, 3 |
395 |
|
GenBank mRNA |
AK012014.1 |
- |
2, 3 |
386 |
|
AK048488.1 |
- |
2, 3 |
418 |
|
AK050391.1 |
- |
2, 3 |
395 |
|
AK078856.1 |
- |
3, 4 |
354 |
|
AK131608.1 |
- |
1, 2 |
306 |
|
AK161812.1 |
- |
2, 3 |
370 |
|
AK163839.1 |
- |
|
390 |
|
AK168006.1 |
- |
2, 3 |
397 |
|
AK168752.1 |
- |
2, 3 |
417 |
|
AK169128.1 |
- |
2, 3 |
396 |
|
AK191398.1 |
- |
1, 2 |
113 |
|
BC005755.1 |
- |
2, 3 |
350 |
2654151 |
View Full Product Details