Gene Transcripts

Gene Symbol
Capsl
Entrez Gene ID
75568
Gene Name
calcyphosine-like
Gene Aliases
1700028N11Rik, AW125205
Chromosome Location
Chr. 15: 9436015 - 9466039 on Build GRCm38
UniGene ID
Mm.82143
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029341.1 NP_083617.2 4 490
GenBank mRNA AK006467.1 - 4 490
BC037621.1 - 3 269
BC061015.1 - 4 448 30297953

View Full Product Details