Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_030559.3 | NP_085036.3 | 17 | 1727 | |
GenBank mRNA | AB056721.1 | - | 17 | 1685 | |
AK032558.1 | - | 8 | 688 | ||
AK049872.1 | - | 17 | 1684 | ||
AK162984.1 | - | 14 | 2050 | ||
AK169694.1 | - | 14 | 2266 | ||
BC003944.1 | - | 5 | 599 | 3487257 | |
BC025626.1 | - | 17 | 1693 | 5320572 |