Gene Transcripts

Gene Symbol
ATXN7
Entrez Gene ID
6314
Gene Name
ataxin 7
Gene Aliases
ADCAII, OPCA3, SCA7
Chromosome Location
Chr. 3: 63864557 - 64003462 on Build GRCh38
UniGene ID
Hs.476595
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000333.3 NP_000324.1 13 3768
NM_001128149.2 NP_001121621.2 9 2948
NM_001177387.1 NP_001170858.1 13 3392
GenBank mRNA AJ000517.1 - 13 3777
AK125125.1 - 1 1268
AL134602.1 - 1 70

View Full Product Details