Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001203.2 | NP_001194.1 | 6 | 578 | |
NM_001256792.1 | NP_001243721.1 | 4 | 474 | ||
NM_001256793.1 | NP_001243722.1 | 4 | 415 | ||
NM_001256794.1 | NP_001243723.1 | 4 | 458 | ||
XM_011532201.2 | XP_011530503.1 | 5 | 457 | ||
XM_017008558.1 | XP_016864047.1 | 6 | 649 | ||
XM_017008559.1 | XP_016864048.1 | 7 | 639 | ||
XM_017008560.1 | XP_016864049.1 | 4 | 531 | ||
XM_017008561.1 | XP_016864050.1 | 5 | 488 | ||
GenBank mRNA | AK293008.1 | - | 4 | 458 | |
AK299930.1 | - | 4 | 415 | ||
AK313642.1 | - | 4 | 474 | ||
BC047773.1 | - | 3 | 374 | 6202951 | |
BC069796.1 | - | 5 | 490 | 7262484 | |
BC069803.1 | - | 5 | 490 | 7262496 | |
D89675.1 | - | 3 | 322 | ||
U89326.1 | - | 6 | 577 |