Gene Transcripts

Gene Symbol
Ubox5
Entrez Gene ID
140629
Gene Name
U box domain containing 5
Gene Aliases
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5
Chromosome Location
Chr. 2: 130589996 - 130630038 on Build GRCm38
UniGene ID
Mm.486003
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001255993.1 NP_001242922.1 7 2737
NM_001255994.1 NP_001242923.1 6 2590
NM_080562.5 NP_542129.2 6 2595
GenBank mRNA AK034114.1 - 6 2544

View Full Product Details