Gene Transcripts

Gene Symbol
Zcchc14
Entrez Gene ID
142682
Gene Name
zinc finger, CCHC domain containing 14
Gene Aliases
AA792890, BDG29
Chromosome Location
Chr. 8: 121598697 - 121652761 on Build GRCm38
UniGene ID
Mm.103202
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080855.2 NP_543131.1 13 4176
GenBank mRNA AB030244.1 - 13 4186
AK035149.1 - 1 922
AK041107.1 - 1 2685
AK122315.1 - 1 3259
BC009020.1 - 2 1954
BC094454.1 - 2 1427
BC132449.1 - 13 4175 40130726
BC137853.1 - 13 4176 8860872

View Full Product Details