Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133733.4 | NP_598494.2 | 6 | 1149 | |
GenBank mRNA | AB040490.1 | - | 6 | 1066 | |
AK018527.1 | - | 1 | 140 | ||
AK028983.1 | - | 6 | 1119 | ||
AK033723.1 | - | 6 | 1151 | ||
AK080481.1 | - | 6 | 1105 | ||
AY259213.1 | - | 7 | 778 | ||
AY326421.1 | - | 6 | 784 | ||
BC026447.1 | - | 6 | 975 | 4459717 |