Gene Transcripts

Gene Symbol
SLC11A1
Entrez Gene ID
6556
Gene Name
solute carrier family 11 member 1
Gene Aliases
LSH, NRAMP, NRAMP1
Chromosome Location
Chr. 2: 218381766 - 218396894 on Build GRCh38
UniGene ID
Hs.591607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000578.3 NP_000569.3 4 717
XM_005246793.3 XP_005246850.1 3 837
XM_005246794.3 XP_005246851.1 4 980
XM_006712709.3 XP_006712772.1 3 837
XM_006712710.3 XP_006712773.1 2 714
XM_006712711.3 XP_006712774.1 2 714
XM_011511684.2 XP_011509986.1 4 980
XM_011511685.2 XP_011509987.1 3 837
XM_017004765.1 XP_016860254.1 3 857
XM_017004766.1 XP_016860255.1 4 948
XM_017004767.1 XP_016860256.1 4 544
GenBank mRNA AB209650.1 - 2 2431
AK294707.1 - 4 458
AK298667.1 - 4 471
AK303398.1 - 4 441
AK307954.1 - 4 717
AK310914.1 - 4 473
AK311405.1 - 3 324
BC033754.1 - 3 305
BC071165.1 - 4 437 5756382
D38171.1 - 2 224
D50402.1 - 4 552
D50403.1 - 4 463
L32185.1 - 4 453

View Full Product Details