Gene Transcripts

Gene Symbol
Slc25a38
Entrez Gene ID
208638
Gene Name
solute carrier family 25, member 38
Gene Aliases
AV019094, BC010801
Chromosome Location
Chr. 9: 120110391 - 120124342 on Build GRCm38
UniGene ID
Mm.236656
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144793.1 NP_659042.1 3, 4 378
GenBank mRNA AK078385.1 - 3, 4 394
BC010801.1 - 3, 4 378 4235245

View Full Product Details