Gene Transcripts
- Gene Symbol
- Alms1
- Entrez Gene ID
- 236266
- Gene Name
- Alstrom syndrome 1
- Gene Aliases
- bbb
- Chromosome Location
- Chr. 6: 85587499 - 85726611 on Build GRCm38
- UniGene ID
- Mm.246967
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145223.2 |
NP_660258.2
|
2, 3 |
621 |
|
GenBank mRNA |
AF425257.1 |
- |
2, 3 |
622 |
|
View Full Product Details