Gene Transcripts

Gene Symbol
Mypop
Entrez Gene ID
232934
Gene Name
Myb-related transcription factor, partner of profilin
Gene Aliases
Myodysa, P42pop
Chromosome Location
Chr. 7: 18990058 - 19001766 on Build GRCm38
UniGene ID
Mm.440069
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145579.3 NP_663554.1 2 477
GenBank mRNA AF364868.1 - 2 256
BC055832.1 - 1 230
CF724206.1 - 2 414
M82903.1 - 1 69

View Full Product Details