Gene Transcripts

Gene Symbol
UGT2B15
Entrez Gene ID
7366
Gene Name
UDP glucuronosyltransferase family 2 member B15
Gene Aliases
HLUG4, UDPGT 2B8, UDPGT2B15, UDPGTH3, UGT2B8
Chromosome Location
Chr. 4: 68646597 - 68670776 on Build GRCh38
UniGene ID
Hs.150207
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001076.3 NP_001067.2 6 1906
GenBank mRNA AF180322.1 - 6 1786
AF548389.1 - 6 1765
BC171879.1 - 2683 9054533
U08854.1 - 6 1769

View Full Product Details