Gene Transcripts

Gene Symbol
Tor2a
Entrez Gene ID
30933
Gene Name
torsin family 2, member A
Gene Aliases
Prosalusin
Chromosome Location
Chr. 2: 32757231 - 32762244 on Build GRCm38
UniGene ID
Mm.200695
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152800.3 NP_690013.1 2, 3 494
GenBank mRNA AK162719.1 - 2, 3 438
AK170979.1 - 1, 2 1673
AK171247.1 - 2, 3 446
BC003466.1 - 2, 3 479 2649388
BC024469.1 - 2, 3 457 4976022
CK031473.1 - 2, 3 355

View Full Product Details