Gene Transcripts

Gene Symbol
Rdh9
Entrez Gene ID
103142
Gene Name
retinol dehydrogenase 9
Gene Aliases
AI121090, Crad3
Chromosome Location
Chr. 10: 127776405 - 127792697 on Build GRCm38
UniGene ID
Mm.235814
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153133.2 NP_694773.1 4 2627
GenBank mRNA AF372838.1 - 4 2627
AK132181.1 - 2698

View Full Product Details