Gene Transcripts

Gene Symbol
Lrrc66
Entrez Gene ID
231296
Gene Name
leucine rich repeat containing 66
Gene Aliases
Chromosome Location
Chr. 5: 73606642 - 73632421 on Build GRCm38
UniGene ID
Mm.168948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153568.1 NP_705796.1 2 476
GenBank mRNA AK051875.1 - 2 483
BC031901.1 - 2 476 5362830

View Full Product Details