Gene Transcripts

Gene Symbol
Lpar4
Entrez Gene ID
78134
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
5730485F04Rik, Gpr23, LPA4, p2y9
Chromosome Location
Chr. X: 106920625 - 106933899 on Build GRCm38
UniGene ID
Mm.90147
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175271.4 NP_780480.2 3 1974
GenBank mRNA AK045289.1 - 3 1944
AK051709.1 - 3 1945
BC052178.1 - 3 1965
BC069178.1 - 3 1884
BC138893.1 - 3 1774 8861915
BC138894.1 - 3 1775 8861916

View Full Product Details