Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175482.3 | NP_780691.2 | 16 | 1914 | |
GenBank mRNA | AK028335.1 | - | 2 | 109 | |
AK036673.1 | - | 6 | 687 | ||
AK052442.1 | - | 8 | 947 | ||
AK129380.1 | - | 6 | 647 | ||
BC066033.1 | - | 16 | 1810 | 6835629 | |
BC088733.1 | - | 16 | 1836 | 30459522 |