Gene Transcripts

Gene Symbol
R3hcc1l
Entrez Gene ID
52013
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
1700036B12Rik, AI450607, C10orf28, D19Ertd386e, GIDRP86, Gidrp88
Chromosome Location
Chr. 19: 42518312 - 42592256 on Build GRCm38
UniGene ID
Mm.254169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177464.4 NP_803415.1 2 283
GenBank mRNA AK082926.1 - 2 284
AK147893.1 - 2 267

View Full Product Details