Gene Transcripts

Gene Symbol
Myo3b
Entrez Gene ID
329421
Gene Name
myosin IIIB
Gene Aliases
A430065P19Rik
Chromosome Location
Chr. 2: 70039029 - 70429195 on Build GRCm38
UniGene ID
Mm.99648
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177376.4 NP_796350.2 35 4804
GenBank mRNA AK040119.1 - 10 1957
AY830392.2 - 34 4776
AY830393.2 - 33 4644

View Full Product Details