Gene Transcripts

Gene Symbol
ALDH3B1
Entrez Gene ID
221
Gene Name
aldehyde dehydrogenase 3 family member B1
Gene Aliases
ALDH4, ALDH7
Chromosome Location
Chr. 11: 68008547 - 68029282 on Build GRCh38
UniGene ID
Hs.523841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000694.3 NP_000685.1 8 1142
NM_001030010.2 NP_001025181.1 7 1031
NM_001161473.2 NP_001154945.1 8 1200
NM_001290058.1 NP_001276987.1 7 791
NM_001290059.1 NP_001276988.1 7 1060
GenBank mRNA AB209651.1 - 6 3297
AK291505.1 - 8 1125
BC013584.2 - 8 1095 3878124
BC014168.2 - 2 126
BC033099.1 - 7 969 5446816
BT009832.1 - 7 1053
CD014083.1 - 6 985
CD014084.1 - 6 716
EF411198.1 - 8 1169
U10868.1 - 8 1100

View Full Product Details